Assay Details
| Gene Symbol: Rabac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873052.3 | XP_004873109.1 | 2 - 3 | 404 | 58 | |
| XM_021250941.1 | XP_021106600.1 | 3 - 4 | 319 | 58 | ||
More Information
Set Membership: |
|

Rabac1
Genomic Map