Assay Details
| Gene Symbol: Tmem241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853159.2 | XP_004853216.1 | 11 - 12 | 686 | 59 | |
| XM_021265154.1 | XP_021120813.1 | 11 - 12 | 686 | 59 | ||
| XM_021265155.1 | XP_021120814.1 | 10 - 11 | 650 | 59 | ||
| XM_021265156.1 | XP_021120815.1 | 9 - 10 | 510 | 59 | ||
| XM_021265157.1 | XP_021120816.1 | 11 - 12 | 686 | 59 | ||
More Information
Set Membership: |
|

Tmem241
Genomic Map