Assay Details
| Gene Symbol: Ciapin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843166.3 | XP_004843223.1 | 9 - 10 | 1203 | 58 | |
| XM_004843167.3 | XP_004843224.1 | 8 - 9 | 985 | 58 | ||
| XM_021257002.1 | XP_021112661.1 | 8 - 9 | 947 | 58 | ||
More Information
Set Membership: |
|

Ciapin1
Genomic Map