Assay Details
Gene Symbol: Rnls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004858346.3 | XP_004858403.1 | 4 - 5 | 799 | 82 | |
XM_004858347.3 | XP_004858404.1 | 4 - 5 | 800 | 82 | ||
XM_004858348.3 | XP_004858405.1 | 2 - 3 | 551 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
