Assay Details
| Gene Symbol: LOC101710247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855011.2 | XP_004855068.1 | 2 - 3 | 503 | 76 | |
| XM_021236533.1 | XP_021092192.1 | 2 - 3 | 368 | 76 | ||
More Information
Set Membership: |
|

LOC101710247
Genomic Map