Assay Details
| Gene Symbol: Ppp1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072018.2 | XP_012927472.2 | 3 | 868 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r2| Gene Symbol: Ppp1r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013072018.2 | XP_012927472.2 | 3 | 868 | 73 | |
Set Membership: |
|