Assay Details
| Gene Symbol: Polr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846488.3 | XP_004846545.1 | 3 - 4 | 349 | 70 | |
| XM_021259770.1 | XP_021115429.1 | 3 - 4 | 349 | 70 | ||
More Information
Set Membership: |
|

Polr1c
Genomic Map