Assay Details
| Gene Symbol: Irf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842752.3 | XP_004842809.1 | 5 - 6 | 617 | 64 | |
| XM_021256622.1 | XP_021112281.1 | 4 - 5 | 496 | 64 | ||
| XM_021256623.1 | XP_021112282.1 | 4 - 5 | 513 | 64 | ||
More Information
Set Membership: |
|

Irf8
Genomic Map