Assay Details
| Gene Symbol: Mrpl58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860627.3 | XP_004860684.1 | 5 - 6 | 550 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl58| Gene Symbol: Mrpl58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860627.3 | XP_004860684.1 | 5 - 6 | 550 | 73 | |
Set Membership: |
|