Assay Details
| Gene Symbol: Nptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855505.3 | XP_004855562.1 | 5 - 6 | 924 | 60 | |
| XM_013076196.2 | XP_012931650.1 | 6 - 7 | 984 | 60 | ||
More Information
Set Membership: |
|

Nptn
Genomic Map