Assay Details
| Gene Symbol: Larp4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854584.3 | XP_004854641.1 | 17 - 18 | 1983 | 60 | |
| XM_021266347.1 | XP_021122006.1 | 16 - 17 | 5252 | 60 | ||
| XM_021266348.1 | XP_021122007.1 | 15 - 16 | 5178 | 60 | ||
| XM_021266349.1 | XP_021122008.1 | 15 - 16 | 5070 | 60 | ||
| XM_021266350.1 | XP_021122009.1 | 14 - 15 | 4943 | 60 | ||
| XM_021266351.1 | XP_021122010.1 | 13 - 14 | 4813 | 60 | ||
| XM_021266353.1 | XP_021122012.1 | 18 - 19 | 2049 | 60 | ||
| XM_021266354.1 | XP_021122013.1 | 17 - 18 | 1988 | 60 | ||
| XM_021266355.1 | XP_021122014.1 | 18 - 19 | 2327 | 60 | ||
| XM_021266356.1 | XP_021122015.1 | 16 - 17 | 1731 | 60 | ||
More Information
Set Membership: |
|

Larp4b
Genomic Map