Assay Details
| Gene Symbol: Hmgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854784.2 | XP_004854841.1 | 1 - 2 | 175 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004854784.2 transcripts.
Set Membership: |
|

Hmgb1
Genomic Map