Assay Details
| Gene Symbol: Arhgap45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865981.3 | XP_004866038.3 | 13 - 14 | 1875 | 58 | |
| XM_004865982.3 | XP_004866039.1 | 13 - 14 | 2912 | 58 | ||
| XM_013065988.2 | XP_012921442.2 | 13 - 14 | 1878 | 58 | ||
| XM_013065989.2 | XP_012921443.1 | 13 - 14 | 2916 | 58 | ||
| XM_013065990.2 | XP_012921444.1 | 12 - 13 | 1482 | 58 | ||
| XM_021244912.1 | XP_021100571.1 | 13 - 14 | 1754 | 58 | ||
| XM_021244913.1 | XP_021100572.1 | 12 - 13 | 1347 | 58 | ||
More Information
Set Membership: |
|

Arhgap45
Genomic Map