Assay Details
| Gene Symbol: Noc4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844024.1 | XP_004844081.1 | 11 - 12 | 1122 | 59 | |
| XM_021257720.1 | XP_021113379.1 | 11 - 12 | 1124 | 59 | ||
| XM_021257721.1 | XP_021113380.1 | 11 - 12 | 1068 | 59 | ||
More Information
Set Membership: |
|

Noc4l
Genomic Map