Assay Details
Gene Symbol: Jph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842763.2 | XP_004842820.1 | 1 - 2 | 547 | 55 | |
XM_021256636.1 | XP_021112295.1 | 1 - 2 | 547 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
