Assay Details
| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864532.3 | XP_004864589.1 | 8 - 9 | 1300 | 58 | |
| XM_004864533.3 | XP_004864590.1 | 8 - 9 | 1292 | 58 | ||
| XM_004864534.3 | XP_004864591.1 | 8 - 9 | 1302 | 58 | ||
| XM_004864535.3 | XP_004864592.1 | 8 - 9 | 1302 | 58 | ||
| XM_013065507.2 | XP_012920961.1 | 8 - 9 | 1295 | 58 | ||
| XM_013065509.2 | XP_012920963.1 | 9 - 10 | 1164 | 58 | ||
| XM_021244282.1 | XP_021099941.1 | 8 - 9 | 1296 | 58 | ||
| XM_021244283.1 | XP_021099942.1 | 9 - 10 | 1160 | 58 | ||
| XM_021244284.1 | XP_021099943.1 | 8 - 9 | 1258 | 58 | ||
| XM_021244285.1 | XP_021099944.1 | 7 - 8 | 1608 | 58 | ||
| XM_021244286.1 | XP_021099945.1 | 5 - 6 | 675 | 58 | ||
| XM_021244287.1 | XP_021099946.1 | 5 - 6 | 672 | 58 | ||
More Information
Set Membership: |
|

Ptprn
Genomic Map