Assay Details
| Gene Symbol: Rpp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838490.2 | XP_004838547.1 | 7 - 8 | 596 | 69 | |
| XM_004838491.2 | XP_004838548.1 | 6 - 7 | 525 | 69 | ||
More Information
Set Membership: |
|

Rpp30
Genomic Map