Assay Details
Gene Symbol: Trim50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840074.3 | XP_004840131.1 | 6 - 7 | 1519 | 57 | |
XM_021254418.1 | XP_021110077.1 | 8 - 9 | 1193 | 57 | ||
XM_021254419.1 | XP_021110078.1 | 5 - 6 | 2125 | 57 | ||
XM_021254421.1 | XP_021110080.1 | 4 - 5 | 2136 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
