Assay Details
| Gene Symbol: Chst15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838760.3 | XP_004838817.1 | 6 - 7 | 2207 | 62 | |
| XM_013069973.2 | XP_012925427.1 | 6 - 7 | 1763 | 62 | ||
| XM_013069974.2 | XP_012925428.1 | 6 - 7 | 1949 | 62 | ||
| XM_013069975.2 | XP_012925429.1 | 6 - 7 | 1484 | 62 | ||
| XM_021253311.1 | XP_021108970.1 | 6 - 7 | 1875 | 62 | ||
| XM_021253312.1 | XP_021108971.1 | 6 - 7 | 2318 | 62 | ||
| XM_021253313.1 | XP_021108972.1 | 6 - 7 | 2425 | 62 | ||
| XM_021253314.1 | XP_021108973.1 | 7 - 8 | 1957 | 62 | ||
| XM_021253315.1 | XP_021108974.1 | 6 - 7 | 1343 | 62 | ||
More Information
Set Membership: |
|

Chst15
Genomic Map