Assay Details
| Gene Symbol: Sts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867342.2 | XP_004867399.1 | 6 - 7 | 1549 | 59 | |
| XM_004867343.2 | XP_004867400.1 | 7 - 8 | 1269 | 59 | ||
| XM_004867344.3 | XP_004867401.1 | 7 - 8 | 1233 | 59 | ||
| XM_004867345.2 | XP_004867402.1 | 6 - 7 | 1113 | 59 | ||
| XM_004867349.3 | XP_004867406.1 | 6 - 7 | 1329 | 59 | ||
| XM_021246399.1 | XP_021102058.1 | 6 - 7 | 1128 | 59 | ||
| XM_021246400.1 | XP_021102059.1 | 7 - 8 | 1120 | 59 | ||
| XM_021246401.1 | XP_021102060.1 | 6 - 7 | 971 | 59 | ||
More Information
Set Membership: |
|

Sts
Genomic Map