Assay Details
| Gene Symbol: LOC101696978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260866.1 | XP_021116525.1 | 7 - 8 | 1361 | 97 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696978| Gene Symbol: LOC101696978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021260866.1 | XP_021116525.1 | 7 - 8 | 1361 | 97 | |