Assay Details
| Gene Symbol: Spock3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874608.3 | XP_004874665.1 | 5 - 6 | 480 | 66 | |
| XM_004874610.3 | XP_004874667.1 | 4 - 5 | 471 | 66 | ||
More Information
Set Membership: |
|

Spock3
Genomic Map