Assay Details
| Gene Symbol: LOC101702684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076264.2 | XP_012931718.1 | 3 - 4 | 603 | 96 | |
| XM_013076267.2 | XP_012931721.1 | 36 - 37 | 5292 | 96 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101702684| Gene Symbol: LOC101702684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076264.2 | XP_012931718.1 | 3 - 4 | 603 | 96 | |
| XM_013076267.2 | XP_012931721.1 | 36 - 37 | 5292 | 96 | ||