Assay Details
| Gene Symbol: Srp54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867567.2 | XP_004867624.1 | 5 - 6 | 609 | 68 | |
| XM_004867568.2 | XP_004867625.1 | 6 - 7 | 744 | 68 | ||
| XM_021246577.1 | XP_021102236.1 | 5 - 6 | 781 | 68 | ||
| XM_021246578.1 | XP_021102237.1 | 5 - 6 | 477 | 68 | ||
More Information
Set Membership: |
|

Srp54
Genomic Map