Assay Details
| Gene Symbol: Fkbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835605.3 | XP_004835662.1 | 6 - 7 | 714 | 71 | |
| XM_004835607.3 | XP_004835664.1 | 5 - 6 | 598 | 71 | ||
| XM_021262758.1 | XP_021118417.1 | 7 - 8 | 714 | 71 | ||
| XM_021262776.1 | XP_021118435.1 | 7 - 8 | 774 | 71 | ||
| XM_021262783.1 | XP_021118442.1 | 6 - 7 | 1439 | 71 | ||
| XM_021262791.1 | XP_021118450.1 | 6 - 7 | 612 | 71 | ||
| XM_021262802.1 | XP_021118461.1 | 6 - 7 | 679 | 71 | ||
More Information
Set Membership: |
|

Fkbp3
Genomic Map