Assay Details
| Gene Symbol: LOC101703656 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837781.3 | XP_004837838.1 | 6 - 7 | 2067 | 77 | |
| XM_021250448.1 | XP_021106107.1 | 6 - 7 | 1794 | 77 | ||
| XM_021250452.1 | XP_021106111.1 | 6 - 7 | 1793 | 77 | ||
More Information
Set Membership: |
|

LOC101703656
Genomic Map