Assay Details
| Gene Symbol: Gprc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855817.3 | XP_004855874.1 | 4 - 5 | 1357 | 89 | |
| XM_013076302.2 | XP_012931756.1 | 4 - 5 | 2376 | 89 | ||
| XM_013076303.2 | XP_012931757.1 | 4 - 5 | 2228 | 89 | ||
| XM_013076304.2 | XP_012931758.1 | 4 - 5 | 1301 | 89 | ||
| XM_021237231.1 | XP_021092890.1 | 3 - 4 | 2680 | 89 | ||
| XM_021237232.1 | XP_021092891.1 | 3 - 4 | 1212 | 89 | ||
More Information
Set Membership: |
|

Gprc5b
Genomic Map