Assay Details
| Gene Symbol: Mtrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021257909.1 | XP_021113568.1 | 9 - 10 | 1475 | 67 | |
| XM_021257910.1 | XP_021113569.1 | 9 - 10 | 1417 | 67 | ||
| XM_021257911.1 | XP_021113570.1 | 9 - 10 | 1321 | 67 | ||
| XM_021257912.1 | XP_021113571.1 | 9 - 10 | 1278 | 67 | ||
More Information
Set Membership: |
|

Mtrf1
Genomic Map