Assay Details
| Gene Symbol: Washc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865313.3 | XP_004865370.1 | 6 - 7 | 890 | 74 | |
| XM_004865314.2 | XP_004865371.1 | 6 - 7 | 890 | 74 | ||
| XM_021245270.1 | XP_021100929.1 | 6 - 7 | 891 | 74 | ||
| XM_021245271.1 | XP_021100930.1 | 8 - 9 | 1163 | 74 | ||
| XM_021245272.1 | XP_021100931.1 | 6 - 7 | 887 | 74 | ||
| XM_021245273.1 | XP_021100932.1 | 6 - 7 | 753 | 74 | ||
| XM_021245274.1 | XP_021100933.1 | 6 - 7 | 793 | 74 | ||
| XM_021245275.1 | XP_021100934.1 | 6 - 7 | 933 | 74 | ||
| XM_021245277.1 | XP_021100936.1 | 6 - 7 | 1881 | 74 | ||
| XM_021245278.1 | XP_021100937.1 | 6 - 7 | 890 | 74 | ||
| XM_021245279.1 | XP_021100938.1 | 6 - 7 | 891 | 74 | ||

Washc1
Genomic Map