Assay Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013076822.2 | XP_012932276.1 | 6 - 7 | 608 | 55 | |
| XM_021238497.1 | XP_021094156.1 | 6 - 7 | 608 | 55 | ||
More Information
Set Membership: |
|

Acadvl
Genomic Map