Assay Details
| Gene Symbol: Tcp11l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852074.2 | XP_004852131.1 | 4 - 5 | 1010 | 69 | |
| XM_021264384.1 | XP_021120043.1 | 4 - 5 | 699 | 69 | ||
| XM_021264385.1 | XP_021120044.1 | 3 - 4 | 394 | 69 | ||
| XM_021264386.1 | XP_021120045.1 | 4 - 5 | 480 | 69 | ||
| XM_021264387.1 | XP_021120046.1 | 4 - 5 | 1011 | 69 | ||
| XM_021264388.1 | XP_021120047.1 | 4 - 5 | 1010 | 69 | ||
More Information
Set Membership: |
|

Tcp11l1
Genomic Map