Assay Details
| Gene Symbol: Raf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870441.3 | XP_004870498.1 | 16 - 17 | 2157 | 55 | |
| XM_004870442.3 | XP_004870499.1 | 16 - 17 | 1939 | 55 | ||
| XM_004870444.3 | XP_004870501.1 | 15 - 16 | 2095 | 55 | ||
| XM_021248807.1 | XP_021104466.1 | 15 - 16 | 1696 | 55 | ||
| XM_021248808.1 | XP_021104467.1 | 15 - 16 | 2044 | 55 | ||
| XM_021248810.1 | XP_021104469.1 | 15 - 16 | 1825 | 55 | ||
More Information
Set Membership: |
|

Raf1
Genomic Map