Assay Details
| Gene Symbol: Dnaaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835352.2 | XP_004835409.1 | 6 - 7 | 982 | 61 | |
| XM_004835353.2 | XP_004835410.1 | 6 - 7 | 983 | 61 | ||
| XM_004835355.2 | XP_004835412.1 | 6 - 7 | 987 | 61 | ||
| XM_013074223.2 | XP_012929677.1 | 6 - 7 | 984 | 61 | ||
| XM_021260798.1 | XP_021116457.1 | 6 - 7 | 990 | 61 | ||
| XM_021260814.1 | XP_021116473.1 | 6 - 7 | 987 | 61 | ||
| XM_021260818.1 | XP_021116477.1 | 6 - 7 | 990 | 61 | ||
| XM_021260820.1 | XP_021116479.1 | 6 - 7 | 987 | 61 | ||
| XM_021260827.1 | XP_021116486.1 | 6 - 7 | 987 | 61 | ||
| XM_021260830.1 | XP_021116489.1 | 6 - 7 | 989 | 61 | ||
More Information
Set Membership: |
|

Dnaaf4
Genomic Map