Assay Details
| Gene Symbol: Dnaaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835352.2 | XP_004835409.1 | 8 - 9 | 1236 | 78 | |
| XM_004835353.2 | XP_004835410.1 | 8 - 9 | 1237 | 78 | ||
| XM_013074223.2 | XP_012929677.1 | 8 - 9 | 1238 | 78 | ||
| XM_021260818.1 | XP_021116477.1 | 9 - 10 | 1545 | 78 | ||
More Information
Set Membership: |
|

Dnaaf4
Genomic Map