Assay Details
| Gene Symbol: LOC110345515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021243099.1 | XP_021098758.1 | 2 - 3 | 325 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021243099.1 transcripts.
Set Membership: |
|

LOC110345515
Genomic Map