Assay Details
| Gene Symbol: Zranb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841444.3 | XP_004841501.1 | 3 - 4 | 423 | 73 | |
| XM_004841445.3 | XP_004841502.1 | 3 - 4 | 423 | 73 | ||
| XM_004841447.3 | XP_004841504.1 | 3 - 4 | 720 | 73 | ||
More Information
Set Membership: |
|

Zranb2
Genomic Map