Assay Details
| Gene Symbol: Vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838934.3 | XP_004838991.1 | 3 - 4 | 401 | 73 | |
| XM_004838935.3 | XP_004838992.1 | 3 - 4 | 401 | 73 | ||
| XM_021253441.1 | XP_021109100.1 | 3 - 4 | 401 | 73 | ||
| XM_021253443.1 | XP_021109102.1 | 3 - 4 | 401 | 73 | ||
| XM_021253444.1 | XP_021109103.1 | 3 - 4 | 401 | 73 | ||
| XM_021253445.1 | XP_021109104.1 | 3 - 4 | 401 | 73 | ||
More Information
Set Membership: |
|

Vti1a
Genomic Map