Assay Details
| Gene Symbol: Mettl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865023.3 | XP_004865080.1 | 9 - 10 | 913 | 61 | |
| XM_021244536.1 | XP_021100195.1 | 9 - 10 | 913 | 61 | ||
More Information
Set Membership: |
|

Mettl17
Genomic Map