Assay Details
| Gene Symbol: Togaram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835615.2 | XP_004835672.1 | 13 - 14 | 4357 | 75 | |
| XM_004835616.2 | XP_004835673.1 | 12 - 13 | 3913 | 75 | ||
| XM_004835619.2 | XP_004835676.1 | 13 - 14 | 4357 | 75 | ||
More Information
Set Membership: |
|

Togaram1
Genomic Map