Assay Details
| Gene Symbol: Togaram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835615.2 | XP_004835672.1 | 17 - 18 | 5006 | 67 | |
| XM_004835616.2 | XP_004835673.1 | 16 - 17 | 4562 | 67 | ||
More Information
Set Membership: |
|

Togaram1
Genomic Map