Assay Details
| Gene Symbol: Vangl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858710.2 | XP_004858767.1 | 6 - 7 | 1568 | 68 | |
| XM_004858711.2 | XP_004858768.1 | 5 - 6 | 1370 | 68 | ||
| XM_021239723.1 | XP_021095382.1 | 6 - 7 | 1485 | 68 | ||
| XM_021239724.1 | XP_021095383.1 | 5 - 6 | 1258 | 68 | ||
More Information
Set Membership: |
|

Vangl2
Genomic Map