Assay Details
| Gene Symbol: Fyb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073529.2 | XP_012928983.1 | 4 - 5 | 1507 | 111 | |
| XM_013073531.2 | XP_012928985.1 | 4 - 5 | 1506 | 111 | ||
| XM_021261473.1 | XP_021117132.1 | 4 - 5 | 1502 | 111 | ||
| XM_021261475.1 | XP_021117134.1 | 4 - 5 | 1499 | 111 | ||
| XM_021261476.1 | XP_021117135.1 | 4 - 5 | 1501 | 111 | ||
More Information
Set Membership: |
|

Fyb1
Genomic Map