Assay Details
| Gene Symbol: Denr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843904.3 | XP_004843961.1 | 7 - 8 | 701 | 73 | |
| XM_021257575.1 | XP_021113234.1 | 7 - 8 | 578 | 73 | ||
| XM_021257576.1 | XP_021113235.1 | 6 - 7 | 578 | 73 | ||
More Information
Set Membership: |
|

Denr
Genomic Map