Assay Details
| Gene Symbol: Ppp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856089.3 | XP_004856146.1 | 2 - 3 | 350 | 63 | |
| XM_004856090.3 | XP_004856147.1 | 2 - 3 | 271 | 63 | ||
| XM_021237437.1 | XP_021093096.1 | 1 - 2 | 459 | 63 | ||
More Information
Set Membership: |
|

Ppp4c
Genomic Map