Assay Details
| Gene Symbol: Stradb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872285.2 | XP_004872342.1 | 9 - 10 | 1061 | 77 | |
| XM_004872286.2 | XP_004872343.1 | 9 - 10 | 1058 | 77 | ||
| XM_013068337.2 | XP_012923791.1 | 9 - 10 | 1061 | 77 | ||
| XM_021250269.1 | XP_021105928.1 | 8 - 9 | 922 | 77 | ||
| XM_021250271.1 | XP_021105930.1 | 7 - 8 | 830 | 77 | ||
More Information
Set Membership: |
|

Stradb
Genomic Map