Assay Details
Gene Symbol: Nsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847401.3 | XP_004847458.1 | 9 - 10 | 2774 | 93 | |
XM_013073021.2 | XP_012928475.1 | 10 - 11 | 2852 | 93 | ||
XM_021260406.1 | XP_021116065.1 | 14 - 15 | 3296 | 93 | ||
XM_021260407.1 | XP_021116066.1 | 12 - 13 | 3081 | 93 | ||
XM_021260408.1 | XP_021116067.1 | 13 - 14 | 3224 | 93 | ||
XM_021260409.1 | XP_021116068.1 | 13 - 14 | 3153 | 93 | ||
XM_021260410.1 | XP_021116069.1 | 14 - 15 | 3413 | 93 | ||
XM_021260411.1 | XP_021116070.1 | 14 - 15 | 3290 | 93 | ||
XM_021260412.1 | XP_021116071.1 | 12 - 13 | 3067 | 93 | ||
XM_021260413.1 | XP_021116072.1 | 10 - 11 | 2063 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
