Assay Details
| Gene Symbol: Otud7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853948.3 | XP_004854005.1 | 5 - 6 | 696 | 62 | |
| XM_004853949.3 | XP_004854006.1 | 5 - 6 | 1038 | 62 | ||
| XM_021265858.1 | XP_021121517.1 | 6 - 7 | 813 | 62 | ||
| XM_021265860.1 | XP_021121519.1 | 3 - 4 | 384 | 62 | ||
| XM_021265861.1 | XP_021121520.1 | 4 - 5 | 670 | 62 | ||
More Information
Set Membership: |
|

Otud7b
Genomic Map