Assay Details
| Gene Symbol: Znf385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868149.2 | XP_004868206.1 | 10 - 11 | 1502 | 65 | |
| XM_004868150.3 | XP_004868207.1 | 9 - 10 | 1465 | 65 | ||
| XM_004868151.3 | XP_004868208.2 | 8 - 9 | 1004 | 65 | ||
| XM_004868152.3 | XP_004868209.1 | 8 - 9 | 1166 | 65 | ||
| XM_004868153.3 | XP_004868210.1 | 8 - 9 | 1101 | 65 | ||
| XM_021247146.1 | XP_021102805.1 | 7 - 8 | 1124 | 65 | ||
| XM_021247147.1 | XP_021102806.1 | 7 - 8 | 1060 | 65 | ||
More Information
Set Membership: |
|

Znf385b
Genomic Map