Assay Details
| Gene Symbol: LOC101700295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851195.3 | XP_004851252.1 | 12 - 13 | 1343 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101700295| Gene Symbol: LOC101700295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004851195.3 | XP_004851252.1 | 12 - 13 | 1343 | 64 | |
Set Membership: |
|