Assay Details
| Gene Symbol: Pigl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870764.3 | XP_004870821.2 | 4 - 5 | 586 | 72 | |
| XM_004870765.3 | XP_004870822.1 | 3 - 4 | 475 | 72 | ||
| XM_021248980.1 | XP_021104639.1 | 4 - 5 | 586 | 72 | ||
| XM_021248981.1 | XP_021104640.1 | 4 - 5 | 586 | 72 | ||
| XM_021248982.1 | XP_021104641.1 | 4 - 5 | 586 | 72 | ||
| XM_021248983.1 | XP_021104642.1 | 4 - 5 | 586 | 72 | ||
| XM_021248984.1 | XP_021104643.1 | 3 - 4 | 475 | 72 | ||
More Information
Set Membership: |
|

Pigl
Genomic Map